MilliporeSigma
Structural characterization of telechelic polyisobutylene diol.
Journal of chromatography. A (2014-12-24)
Sanjib Banerjee, Priyank N Shah, Youncheol Jeong, Taihyun Chang, Kasyap Seethamraju, Rudolf Faust
ABSTRACT
The chemical homogeneity of telechelic polyisobutylene diol (PIB-diol), prepared by hydroboration-oxidation of allyl telechelic PIB obtained by reacting living PIB with allyltrimethylsilane, was investigated by liquid chromatography at critical conditions (LCCC) and HPLC coupled with matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS). A normal phase gradient HPLC method was developed that was able to separate the as-synthesized PIB-diol into three components; PIB-diol, PIB-monool and PIB without any OH functionality. These were analyzed by MALDI-TOF MS, which suggested that the reaction of living PIB with allyltrimethylsilane was incomplete. LCCC using refractive index (RI) detector as a concentration detector allowed separation and quantification of PIB species according to their chemical heterogeneity (PIB-diol=95.3%, PIB-monool=3.3%, non-functional PIB=1.4%). The calculated number average functionality (Fn) of PIB-diol=1.94 suggests high quality of PIB-diol suitable for high molecular weight polyurethane synthesis.
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Dithranol, ≥90% (HPLC)
Dithranol, European Pharmacopoeia (EP) Reference Standard