Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

270474

Sigma-Aldrich

Methanol

≥99.93%, gradient grade, suitable for HPLC

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
41116121
PubChem Substance ID:
grade:
gradient grade
technique(s):
HPLC: suitable
bp:
64.7 °C (lit.)
vapor pressure:
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Product Name

Methanol, suitable for HPLC, gradient grade, 99.93%

grade

gradient grade

Quality Level

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

description

gradient analysis <= 0.005 254nm

Assay

99.93%

form

liquid

autoignition temp.

725 °F

purified by

sub-micron filtration

expl. lim.

36 %

technique(s)

HPLC: suitable

impurities

H2SO4, passes test (darkened)
≤0.001% acetaldehyde
≤0.001% formaldehyde
≤0.1% water

evapn. residue

<0.0003%

color

APHA: ≤10
clear

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

solubility

H2O: passes test

density

0.791 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 203 nm Amax: 1.00
λ: 210 nm Amax: 0.60
λ: 220 nm Amax: 0.30
λ: 230 nm Amax: 0.15
λ: 240 nm Amax: 0.05
λ: 260-400 nm Amax: 0.01

suitability

passes test for HPLC

format

neat

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS specifications.

Other Notes

Contains MnO4- reducers

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Cunha et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 18(6), 1987-1995 (1998-05-15)
ATP analogs substituted in the gamma-phosphorus (ATPgammaS, beta, gamma-imido-ATP, and beta,gamma-methylene-ATP) were used to probe the involvement of P2 receptors in the modulation of synaptic transmission in the hippocampus, because their extracellular catabolism was virtually not detected in CA1 slices.
Ufuk Cobanoglu et al.
Asian Pacific journal of cancer prevention : APJCP, 12(6), 1399-1403 (2011-12-01)
Early diagnosis and prevention is very important for lung cancer patients. Previous studies have emphasized that the level of coenzyme Q10 (CoQ10), present primarily in mitochondria, decreases with age and is low in patients with chronic diseases. Our goal was
Emer P Reeves et al.
Mycopathologia, 158(1), 73-79 (2004-10-19)
Aspergillus fumigatus is a pathogenic fungus capable of causing both allergic lung disease and invasive aspergillosis, a serious, life-threatening condition in neutropenic patients. Aspergilli express an array of mycotoxins and enzymes which may facilitate fungal colonisation of host tissue. In
Robert E Schwartz et al.
The Journal of clinical investigation, 109(10), 1291-1302 (2002-05-22)
We have derived from normal human, mouse, and rat postnatal bone marrow primitive, multipotent adult progenitor cells (MAPCs) that can differentiate into most mesodermal cells and neuroectodermal cells in vitro and into all embryonic lineages in vivo. Here, we show
The conversion of methanol and other O-compounds to hydrocarbons over zeolite catalysts.
Chang CD and Silvestri AJ.
J. Catal., 47(2), 249-259 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service