MilliporeSigma
  • Ezetimibe reduces plant sterol accumulation and favorably increases platelet count in sitosterolemia.
Ezetimibe reduces plant sterol accumulation and favorably increases platelet count in sitosterolemia.
The Journal of pediatrics (2014-12-03)
Rgia A Othman, Semone B Myrie, David Mymin, Louise S Merkens, Jean-Baptiste Roullet, Robert D Steiner, Peter J H Jones
ABSTRACT
To assess if ezetimibe (EZE), a sterol-absorption inhibitor, improves platelet (PLT) count and size relative to its effect on plasma plant sterol (PS) in patients with sitosterolemia (STSL). Patients with STSL (5 males, 3 females, 16-56 years of age) receiving EZE intervention as part of their routine care participated in this study. EZE was discontinued for 14 weeks (off) and then resumed for another 14 weeks (on). Hematology variables along with plasma and red blood cells (RBC) PS and total cholesterol (TC) levels were measured at the end of each phase. EZE increased PLT count (23% ± 9%) and decreased mean PLT volume (MPV; 10% ± 3%, all P < .05). In patients off EZE, PLT counts inversely correlated (r = -0.96 and r = -0.91, all P < .01) with plasma and RBC PS to TC ratio (PS/TC), and MPV positively correlated (r = 0.91, P = .03 and r = 0.93, P = .02) with plasma and RBC PS/TC. EZE reduced plasma and RBC sitosterol (-35% ± 4% and -28% ± 3%), total PS (-37% ± 4% and -28% ± 3%, all P < .0001) levels, and PS/TC (-27% ± 4% and -28% ± 4%, P < .01). EZE reduces plasma and RBC PS levels, while increasing PLT count and decreasing MPV, and thereby may reduce the risk for bleeding in STSL. Plasma PS levels and ABCG5/ABCG8 genes should be analyzed in patients with unexplained hematologic abnormalities.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
5-α-Cholestane, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Stigmasterol, ~95%
Sigma-Aldrich
5-α-Cholestane, ≥97.0% (HPLC)
Beta-sitosterol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
β-Sitosterol, ≥70%
Sigma-Aldrich
Hexane, anhydrous, 95%
Supelco
β-Sitosterol, certified reference material, 100 μg/mL in chloroform
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Supelco
Hexane, analytical standard
Sigma-Aldrich
Campesterol, ~65%
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis