Skip to Content
MilliporeSigma
All Photos(1)

Documents

47124

Supelco

5-α-Cholestane

certified reference material, 10 mg/mL in chloroform

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H48
CAS Number:
Molecular Weight:
372.67
Beilstein:
2051806
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

10 mg/mL in chloroform

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

80-82 °C (lit.)

application(s)

food and beverages

format

single component solution

storage temp.

2-30°C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CCCC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H48/c1-19(2)9-8-10-20(3)23-14-15-24-22-13-12-21-11-6-7-17-26(21,4)25(22)16-18-27(23,24)5/h19-25H,6-18H2,1-5H3/t20-,21-,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

XIIAYQZJNBULGD-XWLABEFZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Rgia A Othman et al.
The Journal of pediatrics, 166(1), 125-131 (2014-12-03)
To assess if ezetimibe (EZE), a sterol-absorption inhibitor, improves platelet (PLT) count and size relative to its effect on plasma plant sterol (PS) in patients with sitosterolemia (STSL). Patients with STSL (5 males, 3 females, 16-56 years of age) receiving
Christophe Regnault et al.
BMC genomics, 15, 666-666 (2014-08-12)
Despite numerous studies suggesting that amphibians are highly sensitive to cumulative anthropogenic stresses, the role pollutants play in the decline of amphibian populations remains unclear. Amongst the most common aquatic contaminants, polycyclic aromatic hydrocarbons (PAHs) have been shown to induce
Simon Hammann et al.
Analytical and bioanalytical chemistry, 407(30), 9019-9028 (2015-10-07)
Triacylglycerols represent the major part (>90%) in most plant oils and have to be eliminated, when the minor compounds such as phytosterols or tocopherols should be analyzed. Here, we used an all liquid-liquid chromatographic technique, countercurrent chromatography (CCC), to fractionate
Jin-Man Kim et al.
Journal of food science, 80(9), C1945-C1951 (2015-08-13)
This study assessed the potential application of gas chromatography (GC) in detecting milk fat (MF) adulteration with vegetable oils and animal fats and of characterizing samples by fat source. One hundred percent pure MF was adulterated with different vegetable oils

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service