Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34859

Sigma-Aldrich

Hexane

≥97.0% (GC), suitable for HPLC, suitable for EPA 1613, EPA ACB B21-02, EPA OTM-45, and EPA ACB B23-05b

Synonym(s):

n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.06
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
69 °C (lit.)
vapor pressure:
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Product Name

Hexane, suitable for HPLC, ≥97.0% (GC)

grade

HPLC grade

Agency

suitable for EPA 1613
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

technique(s)

HPLC: suitable

impurities

≤0.0005% non-volatile matter
≤0.001% free acid (as CH3COOH)
≤0.01% water (Karl Fischer)

evapn. residue

≤0.0005%

transmittance

200 nm, ≥20%
225 nm, ≥80%
250 nm, ≥98%

refractive index

n20/D 1.375 (lit.)

pH

7

bp

69 °C (lit.)

mp

−95 °C (lit.)

density

0.659 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 200 nm Amax: ≤0.70
λ: 225 nm Amax: ≤0.10
λ: 250 nm Amax: ≤0.01

application(s)

food and beverages

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Comparative Study of Gas and Liquid Chromatography Methods for the Determination of Underivatised Neutral and Acidic Cannabinoids and Cholesterol.: This research highlights the essential role of hexane in gas and liquid chromatography for analyzing complex mixtures in pharmaceuticals and clinical diagnostics, demonstrating its critical application in high-precision analytical methods (Czauderna et al., 2024).

  • In Vitro Antioxidant and In Silico Evaluation of the Anti-β-Lactamase Potential of the Extracts of Cylindrospermum alatosporum NR125682 and Loriellopsis cavenicola NR117881.: This study employs hexane to extract bioactive compounds with potential antioxidant and antibacterial activities, showcasing its importance in novel drug discovery and antimicrobial resistance research (Ikhane et al., 2024).

  • Exploration of reducing and stabilizing phytoconstituents in Arisaema dracontium extract for the effective synthesis of Silver nanoparticles and evaluation of their antibacterial and toxicological properties.: Demonstrates the use of hexane in the synthesis of nanoparticles, indicating its utility in nanotechnology and its impact on enhancing antibacterial properties of phytoconstituents (Khattak et al., 2024).

  • Production and Physicochemical Characterization of the Gel Obtained in the Fermentation Process of Blue Corn Flour (Zea mays L.) with Colletotrichum gloeosporioides.: Focuses on the physicochemical properties of gels influenced by hexane extraction, pertinent to food science and material engineering (Villarreal-Rodríguez et al., 2024).

  • Cloning, Expression, Characterization and Immobilization of a Recombinant Carboxylesterase from the Halophilic Archaeon, Halobacterium salinarum NCR-1.: This study involves hexane in the process of enzyme immobilization, highlighting its role in enhancing enzyme stability and activity, crucial for biotechnological applications (Ortega-de la Rosa et al., 2024).


Other Notes

Important notice
  • The article number 34859-4X2.5L will be discontinued. Please order the single bottle 34859-2.5L which is physically identical with the same exact specifications.
  • The article number 34859-6X1L will be discontinued. Please order the single bottle 34859-1L which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao-Tao Zhuang et al.
Nature nanotechnology, 15(3), 192-197 (2020-01-22)
Chirality-the property of an object wherein it is distinguishable from its mirror image-is of widespread interest in chemistry and biology1-6. Regioselective magnetization of one-dimensional semiconductors enables anisotropic magnetism at room temperature, as well as the manipulation of spin polarization-the properties
M A Nurseitova et al.
Journal of dairy science, 102(9), 7723-7733 (2019-07-01)
Milk fat adulteration is a common issue in Central Asia. To assess the current situation in the commercial milk market, 17 milk samples were checked for fatty acid (FA) and sterol profiles to detect potential adulteration using multivariate analysis. Analysis
G Di Nicola et al.
Journal of chromatography. A, 1190(1-2), 120-126 (2008-03-18)
Biodiesel (a mixture of fatty acid esters) is normally analyzed using gas chromatography/flame ionization detection, as specified by the ASTM D6584 and EN14105 standards. This paper proposes a binary gradient method for analyzing biodiesel mixtures using non-aqueous reverse phase HPLC
Comparative evaluation of 11 essential oils of different origin as functional antioxidants, antiradicals and antimicrobials in foods.
Sacchetti G, et al.
Food Chemistry, 91(4), 621-632 (2005)
Ali Mohraz et al.
Langmuir : the ACS journal of surfaces and colloids, 21(12), 5298-5306 (2005-06-01)
The development of model materials and image processing methods to directly visualize and quantify colloidal rod assembly by means of confocal laser scanning microscopy (CLSM) is reported. Monodisperse fluorescent colloidal rods are prepared by the uniaxial extensional deformation of sterically

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service