MilliporeSigma
  • Dose-related effect by maternal exposure to di-(2-ethylhexyl) phthalate plasticizer on inducing hypospadiac male rats.
Dose-related effect by maternal exposure to di-(2-ethylhexyl) phthalate plasticizer on inducing hypospadiac male rats.
Environmental toxicology and pharmacology (2012-12-12)
Mingyong Li, Lin Qiu, Yuanyuan Zhang, Yi Hua, Shengfen Tu, Yun He, Sheng Wen, Qiang Wang, Guanghui Wei
ABSTRACT
The aim of this study was to evaluate dose-related effects on external genitalia of adult male offspring rats by maternal exposure to di-(2-ethylhexyl) phthalate (DEHP) plasticizer. Timed-pregnant rats were given DEHP by gastric intubation at doses of 0, 500, 750 or 1000mg/kg body weight/day from gestation day 12-19 to establish a hypospadiac rat model. The hypospadias was observed and the incidence in three DEHP dosage levels was 10.7%, 30.6% and 37.0%, respectively. With exposed dose increased, mild, moderate and severe hypospadiac rats were distinguished and an increased incidence of severe hypospadias was observed. The other reproductive lesions like reduced penile length and anogenital distance/body weight were observed. The results indicated the dose-related external genitalia teratogenic toxicity, and graded hypospadias on male offspring was resulted from high dosage DEHP maternal exposure.
Sigma-Aldrich
Bis(2-ethylhexyl) phthalate, SAJ first grade, ≥98.0%
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore, for ion-selective electrodes
Supelco
Bis(2-ethylhexyl) phthalate solution, certified reference material, 2000 μg/mL in methanol