Skip to Content
MilliporeSigma

07-3083

Bis(2-ethylhexyl) phthalate

SAJ first grade, ≥98.0%

Synonym(s):

DEHP, Phthalic acid bis(2-ethylhexyl ester), Dioctyl phthalate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
EC Number:
204-211-0
UNSPSC Code:
12352108
PubChem Substance ID:
Beilstein/REAXYS Number:
1890696
MDL number:
Assay:
≥98.0%
Grade:
SAJ first grade
Bp:
386 °C (lit.)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

SAJ first grade

assay

≥98.0%

availability

available only in Japan

dilution

(for analytical testing)

bp

386 °C (lit.)

density

0.985 g/mL at 20 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC

InChI

1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3

InChI key

BJQHLKABXJIVAM-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Bis(2-ethylhexyl) phthalate


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

392.0 °F - Pensky-Martens closed cup

flash_point_c

200 °C - Pensky-Martens closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library