MilliporeSigma
  • Polyaniline/cellulose fiber composite prepared using persulfate as oxidant for Cr(VI)-detoxification.
Polyaniline/cellulose fiber composite prepared using persulfate as oxidant for Cr(VI)-detoxification.
Carbohydrate polymers (2012-12-12)
Xiangyao Liu, Wanpeng Zhou, Xueren Qian, Jing Shen, Xianhui An
ABSTRACT
Cellulose fibers were engineered by in situ oxidative polymerization of aniline using ammonium persulfate as oxidant/initiator. The polyaniline/cellulose fiber composite was used for the treatment of Cr(VI)-contaminated water, and its effect was evaluated. Under the conditions studied, the composite exhibited very high water detoxification efficiency, as a result of reduction of Cr(VI) to Cr(III) in combination with adsorption of the Cr(III) onto the cellulosic substrate. Cellulose fibers used in the study served two purposes simultaneously, i.e., carrier of polyaniline and the adsorbent for Cr(III). The complexation of polyaniline with cellulose fibers provided synergistic effects on Cr(VI)-detoxification.
Product Description
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ≥99.5%
Sigma-Aldrich
Ammonium persulfate, JIS special grade, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Nonne-Apelt′s reagent