Skip to Content
MilliporeSigma

Skip To

09913

Ammonium persulfate

BioUltra, ≥98.0% (RT)

Synonym(s):

AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(NH4)2S2O8
CAS Number:
Molecular Weight:
228.20
PubChem Substance ID:
UNSPSC Code:
41105319
NACRES:
NA.25
EC Number:
231-786-5
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

vapor density

7.9 (vs air)

product line

BioUltra

assay

≥98.0% (RT)

technique(s)

electrophoresis: suitable

impurities

insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected

ign. residue

≤0.05% (as SO4)

pH

1.0-2.5 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤5 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤10 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤500 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

absorption

cut-off at 308 nm in H2O at 1 M

suitability

in accordance for electrophoresis test

SMILES string

N.N.OS(=O)(=O)OOS(O)(=O)=O

InChI

1S/2H3N.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h2*1H3;(H,1,2,3)(H,4,5,6)

InChI key

ROOXNKNUYICQNP-UHFFFAOYSA-N

Application

Ammonium persulfate is a strong oxidizing agent frequently used with tetramethylethylenediamine (TEMED) to catalyze the polymerization of acrylamide in making polyacrylamide gels. Ammonium persulfate is used in a variety of applications that depend upon oxidation and photooxidation reactions.
Catalyst for acrylamide gel polymerization.

Other Notes

Preparation of polyacrylamide gels - polymerization catalyst

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A7460
description

BioUltra, ≥98.0% (RT)

description

BioXtra, ≥98.0%

assay

≥98.0% (RT)

assay

≥98.0%

technique(s)

electrophoresis: suitable

technique(s)

-

grade

Molecular Biology

grade

-

Quality Level

200

Quality Level

200

product line

BioUltra

product line

BioXtra

solubility

H2O: 1 M at 20 °C, clear, colorless

solubility

H2O: 0.5 M at 20 °C, clear, colorless


Still not finding the right product?

Explore all of our products under Ammonium persulfate


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters