Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59845

Supelco

Ammonium sulfate

analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
EC Number:
MDL number:
UNSPSC Code:
85151701
eCl@ss:
38030408
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5%

quality

traceable to NIST SRM 194

shelf life

limited shelf life, expiry date on the label

impurities

21.20%±0.10% total Kjeldahl nitrogen

loss

≤0.1% loss on drying, 110°C

pH

5.0-6.0 (25 °C, 1 M in H2O)

mp

>280 °C (dec.) (lit.)

solubility

soluble, clear, colorless

density

1.77 g/mL at 25 °C (lit.)

format

neat

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium sulfate may be used as a standard solution to determine nitrogen loss in the Kjeldahl analysis which acts as a reference method to determine accurate protein content in dairy products.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Goding.WJ
Monoclonal Antibodies: Principles and Practice, (3) (1996)
Simon Gareth Broadley et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 11), 1450-1459 (2012-10-24)
MshB, a zinc-based deacetylase, catalyses a step in the mycothiol biosynthetic pathway that involves the deacetylation of 1-O-(2-acetamido-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcNAc-Ins), via cleavage of an amide bond, to 1-O-(2-amino-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcN-Ins) and acetate. In this study, MshB was expressed, purified and crystallized. A
Kazunori Sugiura et al.
Biochemical and biophysical research communications, 457(3), 242-248 (2015-01-17)
Intracellular redox state is a critical factor for fundamental cellular functions, including regulation of the activities of various metabolic enzymes as well as ROS production and elimination. Genetically-encoded fluorescent redox sensors, such as roGFP (Hanson, G. T., et al. (2004))
Yohei Y Yamamoto et al.
Journal of molecular biology, 426(14), 2667-2678 (2014-05-27)
Chaperonins are ubiquitous molecular chaperones with the subunit molecular mass of 60kDa. They exist as double-ring oligomers with central cavities. An ATP-dependent conformational change of the cavity induces the folding of an unfolded protein that is captured in the cavity.
James F Hunter et al.
Environmental science & technology, 48(17), 10227-10234 (2014-08-06)
A large number of organic species emitted into the atmosphere contain cycloalkyl groups. While cyclic species are believed to be important secondary organic aerosol (SOA) precursors, the specific role of cyclic moieties (particularly for species with multiple or fused rings)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service