MilliporeSigma
  • Prussian blue caged in spongiform adsorbents using diatomite and carbon nanotubes for elimination of cesium.
Prussian blue caged in spongiform adsorbents using diatomite and carbon nanotubes for elimination of cesium.
Journal of hazardous materials (2012-04-03)
Baiyang Hu, Bunshi Fugetsu, Hongwen Yu, Yoshiteru Abe
ABSTRACT
We developed a spongiform adsorbent that contains Prussian blue, which showed a high capacity for eliminating cesium. An in situ synthesizing approach was used to synthesize Prussian blue inside diatomite cavities. Highly dispersed carbon nanotubes (CNTs) were used to form CNT networks that coated the diatomite to seal in the Prussian blue particles. These ternary (CNT/diatomite/Prussian-blue) composites were mixed with polyurethane (PU) prepolymers to produce a quaternary (PU/CNT/diatomite/Prussian-blue), spongiform adsorbent with an in situ foaming procedure. Prussian blue was permanently immobilized in the cell walls of the spongiform matrix and preferentially adsorbed cesium with a theoretical capacity of 167 mg/g cesium. Cesium was absorbed primarily by an ion-exchange mechanism, and the absorption was accomplished by self-uptake of radioactive water by the quaternary spongiform adsorbent.
Sigma-Aldrich
Diatomaceous earth non-washed, CP
Sigma-Aldrich
Celite® 535, CP
Sigma-Aldrich
Celite® S, filter aid, dried, untreated
Sigma-Aldrich
Diatomaceous earth non-washed, powder
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Celite® 535, filter aid, flux calcined
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Diatomaceous earth
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Hyflo® Super-Cel®, CP
Sigma-Aldrich
Celite® 503, CP
Supelco
Celite® 545 AW, reagent grade
Sigma-Aldrich
Celpure® P65, meets USP/NF testing specifications
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined