MilliporeSigma
  • The combination of dapagliflozin and statins ameliorates renal injury through attenuating the activation of inflammasome-mediated autophagy in insulin-resistant rats.
The combination of dapagliflozin and statins ameliorates renal injury through attenuating the activation of inflammasome-mediated autophagy in insulin-resistant rats.
Journal of biochemical and molecular toxicology (2021-12-24)
Laongdao Thongnak, Nattavadee Pengrattanachot, Sasivimon Promsan, Nichakorn Phengpol, Prempree Sutthasupha, Varanuj Chatsudthipong, Anusorn Lungkaphin
ABSTRACT
Long-term use of a high-fat diet with high-fructose (HFF) intake could promote insulin resistance and induce lipid accumulation leading to kidney injury possibly via impairment of the autophagy process and enhancement of the inflammasome pathway. We investigated whether dapagliflozin as a monotherapy or combined with atorvastatin could restore kidney autophagy impairment and reduce inflammasome activation associated with kidney injury induced by HFF consumption. Male Wistar rats were given an HFF for 16 weeks and then treated with dapagliflozin with or without atorvastatin for 4 weeks. Impaired glucose tolerance, dyslipidemia, renal lipid accumulation along with impaired renal autophagy and activated inflammasome pathway promoted renal injury were exhibited in HFF rats. Dapagliflozin with or without atorvastatin treatment could partially restore disrupted metabolic parameters and reduce kidney injury. In particular, the combination treatment group showed significant amelioration of inflammasome activation and autophagy impairment. In conclusion, the combination therapy of dapagliflozin and atorvastatin has a positive effect on renal injury associated with autophagy and inflammasome activation induced by HFF in insulin-resistant rats. This study is the first report demonstrating the underlying mechanism associated with a combination treatment of dapagliflozin and atorvastatin on autophagy and inflammasome pathways in an insulin-resistant condition. Therefore, dapagliflozin in combination with atorvastatin may be a further preventive or therapeutic strategy for chronic kidney disease in an insulin-resistant or diabetic condition.
Sigma-Aldrich
Anti-Monocyte Chemotactic Protein-1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
IHC Select Secondary Goat Anti-Rabbit IgG Antibody, prediluted, biotinylated, from goat, biotin conjugate
Sigma-Aldrich
Anti-Tumor Necrosis Factor-α Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-GAPDH Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Interleukin-1β Antibody, Chemicon®, from rabbit