Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB1832P

Sigma-Aldrich

Anti-Interleukin-1β Antibody

Chemicon®, from rabbit

Synonym(s):

IL-1beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
neutralization: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

rat ... Il1B(24494)

Specificity

Reacts with rat Interleukin-1beta (IL-1beta).

Immunogen

Highly pure (>98%) recombinant rat IL-1beta.

Application

Anti-Interleukin-1β Antibody is an antibody against Interleukin-1β for use in ELISA, WB, NEUT.
ELISA: 0.5 μg/mL this dilution will allow detection (in direct ELISA) of

0.2-0.4 ng/well when rat IL-1(is used to coat the plate.

Western blot: 0.1-0.2 μg/mL, this dilution should offer detection limit of 1.5-3.0 ng/lane, under reducing or non-reducing conditions.

Neutralization: to yield ½ maximal inhibition (ND50) of the biological activity of rat IL-1beta (0.2 ng/mL), a concentration of 0.09-0.14 μg/mL of antibody is required.

Final working dilutions must be determined by end user.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Linkage

Replaces: 04-462; MABF22

Physical form

Affinity purified Immunoglobulin. Lyophilized from 0.5X PBS pH 7.4, no preservatives. Reconstitute in sterile distilled water.

Storage and Stability

Maintain lyophilized material desiccated below 0°C. After reconstitution maintain at -20°C in convenient undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elina Mäkinen et al.
Brain, behavior, and immunity, 97, 250-259 (2021-07-06)
Increasing evidence shows obesity and poor metabolic health are associated with cognitive deficits, but the mechanistic connections have yet to be resolved. We studied rats selectively bred for low and high intrinsic aerobic capacity in order to test the association
A highly conserved Toxo1 haplotype directs resistance to toxoplasmosis and its associated caspase-1 dependent killing of parasite and host macrophage.
Cavailles, P; Flori, P; Papapietro, O; Bisanz, C; Lagrange, D; Pilloux, L; Massera et al.
PLoS Pathogens null
Xiaonan Zhuang et al.
Journal of cellular and molecular medicine, 25(1), 147-160 (2020-11-19)
We investigated how Src-homology 2-domain phosphatase-1 (SHP-1) regulates the inflammatory response in endotoxin-induced uveitis (EIU), and the signalling pathways involved. One week after intravitreal injection of short hairpin RNA targeting SHP-1 or SHP-1 overexpression lentivirus in rats, we induced ocular
Erika Hernández-Aquino et al.
World journal of gastroenterology, 23(24), 4354-4368 (2017-07-15)
To study the molecular mechanisms involved in the hepatoprotective effects of naringenin (NAR) on carbon tetrachloride (CCl4)-induced liver fibrosis. Thirty-two male Wistar rats (120-150 g) were randomly divided into four groups: (1) a control group (n = 8) that received
Qiong Cao et al.
PloS one, 12(10), e0186764-e0186764 (2017-10-20)
Microglia activation and associated inflammatory response are involved in the pathogenesis of different neurodegenerative diseases. We have reported that Notch-1 and NF-κB/p65 signalling pathways operate in synergy in regulating the production of proinflammatory mediators in activated microglia. In the latter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service