MilliporeSigma
  • Unveiling the Crucial Role of Type IV Secretion System and Motility of Helicobacter pylori in IL-1β Production via NLRP3 Inflammasome Activation in Neutrophils.
Unveiling the Crucial Role of Type IV Secretion System and Motility of Helicobacter pylori in IL-1β Production via NLRP3 Inflammasome Activation in Neutrophils.
Frontiers in immunology (2020-06-26)
Ah-Ra Jang, Min-Jung Kang, Jeong-Ih Shin, Soon-Wook Kwon, Ji-Yeon Park, Jae-Hun Ahn, Tae-Sung Lee, Dong-Yeon Kim, Bo-Gwon Choi, Myoung-Won Seo, Soo-Jin Yang, Min-Kyoung Shin, Jong-Hwan Park
ABSTRACT
Helicobacter pylori is a gram-negative, microaerophilic, and spiral-shaped bacterium and causes gastrointestinal diseases in human. IL-1β is a representative cytokine produced in innate immune cells and is considered to be a key factor in the development of gastrointestinal malignancies. However, the mechanism of IL-1β production by neutrophils during H. pylori infection is still unknown. We designed this study to identify host and bacterial factors involved in regulation of H. pylori-induced IL-1β production in neutrophils. We found that H. pylori-induced IL-1β production is abolished in NLRP3-, ASC-, and caspase-1/11-deficient neutrophils, suggesting essential role for NLRP3 inflammasome in IL-1β response against H. pylori. Host TLR2, but not TLR4 and Nod2, was also required for transcription of NLRP3 and IL-1β as well as secretion of IL-1β. H. pylori lacking cagL, a key component of the type IV secretion system (T4SS), induced less IL-1β production in neutrophils than did its isogenic WT strain, whereas vacA and ureA were dispensable. Moreover, T4SS was involved in caspase-1 activation and IL-1β maturation in H. pylori-infected neutrophils. We also found that FlaA is essential for H. pylori-mediated IL-1β production in neutrophils, but not dendritic cells. TLR5 and NLRC4 were not required for H. pylori-induced IL-1β production in neutrophils. Instead, bacterial motility is essential for the production of IL-1β in response to H. pylori. In conclusion, our study shows that host TLR2 and NLRP3 inflammasome and bacterial T4SS and motility are essential factors for IL-1β production by neutrophils in response to H. pylori.
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Millipore
Thioglycollate Broth (USP Alternative), suitable for microbiology, NutriSelect® Plus
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Sigma-Aldrich
Histopaque®-1119, sterile-filtered, density: 1.119
Sigma-Aldrich
Glyburide, meets USP testing specifications
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Caspase-1 Inhibitor II, The Caspase-1 Inhibitor II, also referenced under CAS 178603-78-6, controls the biological activity of Caspase-1. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Nystatin, ≥4,400 units/mg
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥900 μg per mg (as vancomycin base)
Sigma-Aldrich
Triton X-100, BioXtra