MilliporeSigma
  • Pathological consequences of anti-citrullinated protein antibodies in tear fluid and therapeutic potential of pooled human immune globulin-eye drops in dry eye disease.
Pathological consequences of anti-citrullinated protein antibodies in tear fluid and therapeutic potential of pooled human immune globulin-eye drops in dry eye disease.
The ocular surface (2019-10-14)
Jieun Kwon, Bayasgalan Surenkhuu, Ilangovan Raju, Nour Atassi, Jessica Mun, Yi-Fan Chen, Monazzah Akbar Sarwar, Mark Rosenblatt, Anubhav Pradeep, Seungwon An, Nikhil Dhall, Christine Mun, Sandeep Jain
ABSTRACT
To investigate the role of Anti-Citrullinated Protein autoantibodies (ACPAs) in the pathology of dry eye disease (DED) and the therapeutic potential of pooled human immune globulin-eye drops in these patients. We investigated the presence of citrullinated proteins and ACPAs in ocular surface wash (OSW) and conjunctival impressions from patients with DED and determined the pathological consequences of OSW with high ACPA using in vitro experiments and in vivo murine models. We performed a randomized, double-masked, pilot clinical trial to determine the safety, tolerability and preliminary efficacy of using pooled human immune globulin-eye drops to treat DED patients with ACPAs in OSW. We found that neutrophils are a source of citrullinated proteins on the ocular surface of DED patients. We detected significantly higher immunoglobulin amount and presence of several species of ACPAs in OSW from DED patients. We also found that OSW with high ACPA contributes to production of NETs, and that ACPAs cause ocular surface disease in murine eyes, both of which are reduced with addition of Immune globulins. As compared to Vehicle treatment, pooled human immune globulin-eye drops (IVIG 4 mg/mL) twice a day for 8 weeks caused significant reduction in signs and symptoms of DED with no difference in tolerability or adverse events. This is the first report demonstrating ACPAs in OSW of DED patients and their contribution to ocular surface disease. The first-in-human clinical trial suggests that pooled immune globulin-eye drops are a potential new class of biologic therapies for Dry Eye patients.
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Sigma-Aldrich
Mouse IgA ELISA Kit
Millipore
Millicell® EZ Slide, 4-well glass chamber slide, 1.7 cm2/well, 16/pk, sterile
Millipore
MILLIPLEX® Human Autoimmune Autoantibody Panel, Milliplex®