Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PEZGS0416

Millipore

Millicell® EZ Slide

4-well glass chamber slide, 1.7 cm2/well, 16/pk, sterile

Synonym(s):

4 well glass slide, Chamber slides

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.22

material

polypropylene wells
polystyrene lid
soda-lime glass slide

Quality Level

sterility

ethylene oxide treated
sterile

feature

Raised corner ridges on the lid facilitate stacking of multiple units

packaging

box of 16 (4 trays, 4 units/tray)

manufacturer/tradename

Millicell®

technique(s)

cell based assay: suitable
cell culture | mammalian: suitable
immunocytochemistry: suitable

L × W × H

75 mm × 25 mm × 1 mm, slide only
77.8 mm × 33.8 mm × 13.9, excluding lid

L × W

77.8 mm × 33.8 mm × 16.2 mm, including lid

size

4 wells

well surface area

1.7 cm2

well volume

500-1700 μL

shipped in

ambient

General description

Millicell® EZ Slide allows the user to grow, fix, and stain cells directly on a microscope slide. There is no need to disassemble the unit or transfer cells from one device to another prior to analysis. The polypropylene wells are resistant to most fixatives used, so the cells can be grown on the slide and stained right in the well. Once stained, the microscope slide can be easily removed by pressing down the four side tabs of the unit and lifting the wells off of the slide. This easy slide removal reduces the risk of damaging the cells and no special tools are required to remove the wells from the slide. Raised corner ridges on the cover facilitate stacking of multiple units in the incubator, or they can be incubated in the tray supplied with the units.

Preparation Note

  1. Culture, fix, and stain the cells according to preferred protocol.
  2. To access the microscope slide, hold the Millicell® EZ SLIDE down on a flat surface and break the four tabs by pushing them down with the thumb. Remove the cover. Hold the slide down and lift the wells upward to remove. Remove slide from holder.

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jieun Kwon et al.
The ocular surface, 18(1), 80-97 (2019-10-14)
To investigate the role of Anti-Citrullinated Protein autoantibodies (ACPAs) in the pathology of dry eye disease (DED) and the therapeutic potential of pooled human immune globulin-eye drops in these patients. We investigated the presence of citrullinated proteins and ACPAs in
Timothy J Davies et al.
Stem cells and development, 21(10), 1688-1700 (2011-09-22)
The derivation of pluripotent embryonic stem cells (ESCs) from a variety of genetic backgrounds remains a desirable objective in the generation of mice functionally deficient in genes of interest and the modeling of human disease. Nevertheless, disparity in the ease
Xinfang Yu et al.
Journal of hematology & oncology, 13(1), 40-40 (2020-05-03)
Aberrant activation of DNA damage response (DDR) is a major cause of chemoresistance in colorectal cancer (CRC). CHK1 is upregulated in CRC and contributes to therapeutic resistance. We investigated the upstream signaling pathways governing CHK1 activation in CRC. We identified

Articles

High titer lentiviral particles for LC3 variants used for live cell analysis of cellular autophagy.

High titer lentiviral particles including beta-actin, alpha-tubulin and vimentin used for live cell analysis of cytoskeleton structure proteins.

High titer lentiviral particles including beta-actin, alpha-tubulin and vimentin used for live cell analysis of cytoskeleton structure proteins.

High titer lentiviral particles including beta-actin, alpha-tubulin and vimentin used for live cell analysis of cytoskeleton structure proteins.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service