Skip to Content
MilliporeSigma
All Photos(3)

Documents

HAIAB-10K

Millipore

MILLIPLEX® Human Autoimmune Autoantibody Panel

Milliplex®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

species reactivity

human

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

General description

Autoantibody formation is principal to the pathogenesis of a variety of autoimmune diseases. Dysregulated apoptosis and the subsequent defective clearance of cellular debris leads to the exposure of autoantigens and the generation of autoantibodies. The presence of autoantibodies may indicate disease activity, prognosis, and clinical associations related to a variety of autoimmune diseases. These autoimmune diseases include, but are not limited to, Sjögren′s Syndrome, Systemic Lupus Erythematosus, Mixed Connective Tissue Disease, Systemic Sclerosis, CREST syndrome, Polymyositis/Dermatomyositis, Vasculitis, Anti-Phospholipid Syndrome, and various overlap syndromes of these diseases.

MILLIPLEX® Human Autoimmune Autoantibody Bead Panel is a 20-plex kit to be used for the simultaneous detection and profiling of any or all of the following analytes in serum and plasma samples: Anti-C1q, Anti-Centromere Protein A (CENP-A), Anti-Centromere Protein B (CENP-B), Anti-β 2-Glycoprotein, Anti-Jo-1, Anti-Ku, Anti-Mi-2, Anti-Myeloperoxidase, Anti-Proliferating cell nuclear antigen A (PCNA), Anti-Alanyl-tRNA synthetase (PL-12), Anti-PM/Scl-100, Anti-Proteinase 3, Anti-Ribosomal P, Anti-Ribonucleoprotein (RNP), Anti-RNP/Smith (RNP/Sm), Anti-Scl-70, Anti-Sm, Anti-Sjögren′s Syndrome-related antigen B/La (SSB/La), Anti-Anti-Sjögren′s Syndrome-related antigen A/Ro52 kDa (SSA/Ro52), Anti-Sjögren′s Syndrome-related antigen A/Ro60 kDa (SSA/Ro60). This panel includes 4 assay control beads as part of the base kit format. From our single to multiplex biomarker solutions, we partner with you to design, develop, analytically verify, and build the most comprehensive library available for autoantibody detection.

Panel Type: Immune Response

Specificity

Cross Reactivty
Samples were incubated with no antigen, irrelevant antigen, or specific antigen. Incubation with a partcular antigen led to selective abrogation of the reactivity, while reactivity was unaffected by the irrelevant antigen.

Application

  • Analytes: β2-Glycoprotein, C1q, CENP-B (Centromere Protein B), CENP-A (Centromere Protein A), Jo-1, Ku, Mi-2, Myeloperoxidase (MPO), PCNA (Proliferating Cell Nuclear Antigen A), PL-12 (Alanyl-tRNA synthetase), PM/Scl 100, Proteinase 3, RNP (Ribonucleoprotein), RNP/Smith (RNP/Sm), Ribosomal P, Scl-70, Sm, SSB/La (Sjögren′s Syndrome-related antigen B/La), SSA/Ro60 (Sjögren′s Syndrome-related antigen A/Ro60 kDa), SSA/Ro52 (Sjögren′s Syndrome-related antigen A/Ro52 kDa)
  • This is a qualitative assay.
  • Four assay control beads are included in the base kit.
  • Recommended Sample type: serum and plasma.
  • Assay Run Time: Overnight
  • Research Category: Category Inflammation & Immunology
  • Research Subcategory: Autoimmune disease, Inflammation, Immune Response, Immune Disorders

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

96 well plate
Everything you need in a single kit.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Li et al.
JCI insight, 5(2) (2019-12-27)
Genetic variants within or near the interferon regulatory factor 5 (IRF5) locus associate with systemic lupus erythematosus (SLE) across ancestral groups. The major IRF5-SLE risk haplotype is common across populations, yet immune functions for the risk haplotype are undefined. We
Jieun Kwon et al.
The ocular surface, 18(1), 80-97 (2019-10-14)
To investigate the role of Anti-Citrullinated Protein autoantibodies (ACPAs) in the pathology of dry eye disease (DED) and the therapeutic potential of pooled human immune globulin-eye drops in these patients. We investigated the presence of citrullinated proteins and ACPAs in

Related Content

Multiplex autoantibody assays detect cytokine, autoimmune, and cancer autoantibodies, advancing research comprehensively.

Multiplex autoantibody assays detect cytokine, autoimmune, and cancer autoantibodies, advancing research comprehensively.

Multiplex autoantibody assays detect cytokine, autoimmune, and cancer autoantibodies, advancing research comprehensively.

Multiplex autoantibody assays detect cytokine, autoimmune, and cancer autoantibodies, advancing research comprehensively.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service