Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.93600

Supelco

Silver nitrate

EMPLURA®

Synonym(s):

Silver nitrate, Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352302
grade:
laboratory grade
form:
solid
Pricing and availability is not currently available.

grade

laboratory grade

vapor density

5.8 (vs air)

product line

EMPLURA®

form

solid

mp

212 °C (dec.) (lit.)

density

4.350 g/cm3

storage temp.

room temp

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.93600.0027: 25 g, Minimum order quantity: 20
1.93600.0121: 100 g, Minimum order quantity: 20
1.93600.0521: 500 g, Minimum order quantity: 20

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service