Skip to Content
MilliporeSigma
Micro particle size standard based on polystyrene monodisperse analytical standard, size: 6.0 μm

Micro particle size standard based on polystyrene monodisperse

Micro particle size standard based on polystyrene monodisperse analytical standard, size: 6.0 μm
Synonym(s):
Size standards

Products

Brand
Physical Form
Format
Quality Segment
Application
Particle Size (µm)
Reference Material Type
Special Grade
Showing 1-15 of 15
Compare
Product Number
Description
Pricing