Skip to Content
MilliporeSigma
All Photos(1)

Documents

1711133

USP

Lypressin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

[Lys8]-Vasopressin, Lysine vasopressin, Lyspressin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H65N13O12S2
CAS Number:
Molecular Weight:
1056.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lypressin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CC(O)=O.NCCCCC(NC(=O)C1CCCN1C(=O)C2CSSCC(N)C(=O)NC(Cc3ccc(O)cc3)C(=O)NC(Cc4ccccc4)C(=O)NC(CCC(N)=O)C(=O)NC(CC(N)=O)C(=O)N2)C(=O)NCC(N)=O

InChI

1S/C46H65N13O12S2.C2H4O2/c47-17-5-4-9-29(40(65)52-22-38(51)63)54-45(70)35-10-6-18-59(35)46(71)34-24-73-72-23-28(48)39(64)55-31(20-26-11-13-27(60)14-12-26)43(68)56-32(19-25-7-2-1-3-8-25)42(67)53-30(15-16-36(49)61)41(66)57-33(21-37(50)62)44(69)58-34;1-2(3)4/h1-3,7-8,11-14,28-35,60H,4-6,9-10,15-24,47-48H2,(H2,49,61)(H2,50,62)(H2,51,63)(H,52,65)(H,53,67)(H,54,70)(H,55,64)(H,56,68)(H,57,66)(H,58,69);1H3,(H,3,4)

InChI key

NKWAIKZITYLOMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Lys-Gly-NH2 [Disulfide Bridge: 1-6]

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lypressin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monograph such as Vasopressin.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service