Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1477615

USP

Octreotide Non-Cyclic System Suitability Marker

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-Phenylalanyl-L-cysteinyl-L-phenylalanyl-D-tryptophyl-L-lysyl-L-threonyl-S-[(acetylamino)methyl]-N-[(1R,2R)-2-hydroxy-1-(hydroxymethyl)propyl]-L-cysteinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H73N11O11S2
CAS Number:
Molecular Weight:
1092.33
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

octreotide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

General description

Octreotide Non-Cyclic System Suitability Marker USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Octreotide Non-Cyclic System Suitability Marker USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Octreotide Acetate

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Octreotide Acetate
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(4) , 3228-3228 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service