Skip to Content
MilliporeSigma
All Photos(1)

Documents

1585017

USP

Pyrazinamide Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Pyrazinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N2O2
CAS Number:
Molecular Weight:
124.10
Beilstein:
112305
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

API family

pyrazinamide

packaging

pkg of 25 mg

manufacturer/tradename

USP

mp

222-225 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecules)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)c1cnccn1

InChI

1S/C5H4N2O2/c8-5(9)4-3-6-1-2-7-4/h1-3H,(H,8,9)

InChI key

NIPZZXUFJPQHNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Pyrazinamide Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Pyrazinamide

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service