Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.18305

Supelco

Hexane-1-sulfonic acid sodium salt

for ion pair chromatography LiChropur

Synonym(s):

Hexane-1-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NaO3S
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
12352107
EC Index Number:
220-601-3
NACRES:
NB.21

Quality Level

Assay

≥99% (acidimetric)

technique(s)

ion pair chromatography: suitable

loss

≤2 loss on drying, 120°C; 4h; vacuum

transmittance

200 nm, ≥70%
220 nm, ≥90%
250 nm, ≥98%
(0.005 M; 1 cm in H2O)

pH

5.5-7.5 (20 °C, 100 g/L in H2O)

suitability

passes test for identity (IR)

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])CCCCCC

InChI

1S/C6H14O3S.Na/c1-2-3-4-5-6-10(7,8)9;/h2-6H2,1H3,(H,7,8,9);/q;+1/p-1

InChI key

QWSZRRAAFHGKCH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Hexane-1-sulfonicacid sodium salt is a commonly used ion-pair reagent in ion-pairchromatography. It is used as an ion-pair reagent to generate neutralequilibrium with the analytes, leading to increased retention and improvedseparation during the chromatographic analysis.

Application

Hexane-1-sulfonic acid sodium salt can be used as a:
  • Part of the mobile phase composition in an HPLC stability-indicating method for the simultaneous determination of water-soluble vitamins and preservatives.
  • Buffer component in a reverse-phase ultra-performance liquid chromatography (UPLC) method for the simultaneous determination of Sitagliptin phosphate monohydrate and Metformin hydrochloride in pharmaceutical dosage forms.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of some water-soluble vitamins and preservatives in multivitamin syrup by validated stability-indicating high-performance liquid chromatography method
Vidovic S, et al.
Journal of Chromatography A, 1202(2) (2008)
Simultaneous determination of sitagliptin phosphate monohydrate and metformin hydrochloride in tablets by a validated UPLC method
Malleswararao CSN, et al.
Scientia Pharmaceutica, 80(1), 139-152 (2012)

Articles

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service