Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1500273

USP

Paroxetine Related Compound F

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-Methylparoxetine, (3S-trans)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-1-methylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22FNO3
CAS Number:
Molecular Weight:
343.39
Beilstein:
7468178
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

paroxetine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1CC[C@H]([C@H](COc2ccc3OCOc3c2)C1)c4ccc(F)cc4

InChI

1S/C20H22FNO3/c1-22-9-8-18(14-2-4-16(21)5-3-14)15(11-22)12-23-17-6-7-19-20(10-17)25-13-24-19/h2-7,10,15,18H,8-9,11-13H2,1H3/t15-,18-/m0/s1

InChI key

MOJZPKOBKCXNKG-YJBOKZPZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Paroxetine Related Compound F USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Paroxetine Hydrochloride
  • Paroxetine Extended-Release Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Terje Vasskog et al.
Waste management (New York, N.Y.), 29(11), 2808-2815 (2009-07-15)
Sewage and sewage sludge is known to contain pharmaceuticals, and since sewage sludge is often used as fertilizer within agriculture, the reduction of the selective serotonin reuptake inhibitors (SSRIs) Citalopram, Sertraline, Paroxetine, Fluvoxamine and Fluoxetine during composting has been investigated.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service