Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000579

Paroxetine hydrochloride (anhydrous) impurity C

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(3S,4R)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-1-benzyl-4-(4-fluorophenyl)piperidine, N-Benzylparoxetine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26FNO3
CAS Number:
Molecular Weight:
419.49
Beilstein:
9809201
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

paroxetine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

c1ccccc1.Fc2ccc(cc2)[C@@H]3CCN(C[C@H]3COc4ccc5OCOc5c4)Cc6ccccc6

InChI

1S/C26H26FNO3/c27-22-8-6-20(7-9-22)24-12-13-28(15-19-4-2-1-3-5-19)16-21(24)17-29-23-10-11-25-26(14-23)31-18-30-25/h1-11,14,21,24H,12-13,15-18H2/t21-,24-/m0/s1

InChI key

AGTKMLMRHVWXHN-URXFXBBRSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Paroxetine hydrochloride (anhydrous) impurity C EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service