Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1140393

USP

Clonidine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-(2,6-Dichloroanilino)-2-imidazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9Cl2N3
CAS Number:
Molecular Weight:
230.09
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clonidine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Clc1c(c(ccc1)Cl)NC2=NCCN2

InChI

1S/C9H9Cl2N3/c10-6-2-1-3-7(11)8(6)14-9-12-4-5-13-9/h1-3H,4-5H2,(H2,12,13,14)

InChI key

GJSURZIOUXUGAL-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application


  • Pharmacological research in hypertension: Clonidine, as an alpha-2 adrenergic agonist, is widely researched for its efficacy in controlling high blood pressure, highlighting its role in cardiovascular studies and its impact on hypertension management (Luo et al., 2023).

  • Neuroscience studies on ADHD: Studies utilizing clonidine in the treatment of Attention Deficit Hyperactivity Disorder (ADHD) emphasize its effectiveness as a non-stimulant option, contributing to neuroscience research by exploring its neuromodulatory effects (Goodwin et al., 2023).

  • Opioid withdrawal management: Clonidine′s application in mitigating opioid withdrawal symptoms underlines its critical role in addiction medicine, providing a pharmacological approach to support recovery processes (Riccardi et al., 2023).

  • Preclinical cardiovascular research: The utilization of clonidine in preclinical studies involves detailed cardiovascular assessments, helping to elucidate the drug′s mechanism in blood pressure regulation and heart rate control, crucial for developing targeted therapies (Salim et al., 2022).

  • In vitro pharmacological profiling: Clonidine is extensively used in in vitro studies for pharmacological profiling, helping to determine its interaction with various neurotransmitter systems and its pharmacodynamics (Flinspach et al., 2022).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service