Skip to Content
MilliporeSigma

Skip To

1140247

USP

Clomipramine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-Chloro-10,11-dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine hydrochloride, Anafranil hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H23ClN2 · HCl
CAS Number:
Molecular Weight:
351.31
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

clomipramine

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN(C)CCCN1c2ccccc2CCc3ccc(Cl)cc13

InChI

1S/C19H23ClN2.ClH/c1-21(2)12-5-13-22-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)22;/h3-4,6-7,10-11,14H,5,8-9,12-13H2,1-2H3;1H

InChI key

WIMWMKZEIBHDTH-UHFFFAOYSA-N

Gene Information

human ... SLC6A4(6532)

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clomipramine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Clomipramine Compounded Oral Suspension, Veterinary
  • Clomipramine Hydrochloride
  • Clomipramine Hydrochloride Capsules

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C2360000BP980C7291
description

United States Pharmacopeia (USP) Reference Standard

description

for microbiological assay, European Pharmacopoeia (EP) Reference Standard

description

British Pharmacopoeia (BP) Reference Standard

description

≥98% (HPLC), powder

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

BP

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

-

Gene Information

human ... SLC6A4(6532)

Gene Information

human ... SLC6A4(6532)

Gene Information

human ... SLC6A4(6532)

Gene Information

human ... HTR1A(3350), HTR1B(3351), HTR1D(3352), HTR1E(3354), HTR1F(3355), HTR2A(3356), HTR2B(3357), HTR2C(3358), HTR3A(3359), HTR3B(9177), HTR3C(170572), HTR3D(200909), HTR3E(285242), HTR4(3360), HTR5A(3361), HTR5B(645694), HTR6(3362), HTR7(3363), SLC6A4(6532)

agency

USP

agency

-

agency

-

agency

-


Still not finding the right product?

Explore all of our products under Clomipramine hydrochloride


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Central nervous system,Cardiovascular

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters