Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1036959

USP

Anise oil

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Anise seed oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
629884
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

API family

anise oil

manufacturer/tradename

USP

refractive index

n20/D 1.554 (lit.)

bp

232 °C (lit.)

density

0.980 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Anise oil USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service