Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10521

Sigma-Aldrich

Anise oil

nature identical

Synonym(s):

Anise seed oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
629884
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.25

Quality Level

refractive index

n20/D 1.552-1.561
n20/D 1.554 (lit.)

bp

232 °C (lit.)

density

0.980 g/mL at 25 °C (lit.)
0.978-0.990 (D20/4 )

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Christopher J Derrick et al.
Cardiovascular research (2021-02-23)
Vertebrate heart development requires the complex morphogenesis of a linear tube to form the mature organ, a process essential for correct cardiac form and function requiring coordination of embryonic laterality, cardiac growth, and regionalised cellular changes. While previous studies have
Cong Huang et al.
Immunity, 48(5), 1006-1013 (2018-05-17)
Tissue damage and infection are deemed likewise triggers of innate immune responses. But whereas neutrophil responses to microbes are generally protective, neutrophil recruitment into damaged tissues without infection is deleterious. Why neutrophils respond to tissue damage and not just to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service