Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z777031

Zenix® SEC-300 (3 µm) HPLC Columns

L x I.D. 20 cm × 7.8 mm, hydrophilic polymer phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

Product Name

Zenix® SEC-300 Gel Filtration Column, 3 μm, L × I.D. 20 cm × 7.8 mm, 3 μm particle size, stainless steel

material

stainless steel

Agency

suitable for USP L59

product line

Zenix®

mol wt

5,000-1,250,000

manufacturer/tradename

Sepax 213300-7820

packaging

1 ea of

parameter

20 Mpa max. pressure (3000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

20 cm × 7.8 mm

impurities

<10 ppm metals

matrix

spherical silica
fully porous particle

matrix active group

hydrophilic polymer phase

particle size

3 μm

pore size

300 Å

operating pH

2.0-8.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Like their 5 micron siblings, 3 micron Zenix SEC columns feature a hydrophilic surface layer to prevent the sample from interacting with the underlying silica. Select 100, 300 or 500 A pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.

Zenix User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Zenix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service