Skip to Content
MilliporeSigma
All Photos(1)

Documents

Y0002161

Nomegestrol acetate for ID and assay CRS

CRS, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Nomegestrol acetate, 17-Acetoxy-6-methyl-19-norpregna-4,6-diene-3,20-dione, NOMAc, Org 10486-0

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30O4
CAS Number:
Molecular Weight:
370.48
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

API family

megesterol

manufacturer/tradename

EDQM

storage temp.

2-8°C

SMILES string

CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C(C)=O

InChI

1S/C23H30O4/c1-13-11-20-18(17-6-5-16(26)12-19(13)17)7-9-22(4)21(20)8-10-23(22,14(2)24)27-15(3)25/h11-12,17-18,20-21H,5-10H2,1-4H3/t17-,18-,20-,21+,22+,23+/m1/s1

InChI key

IIVBFTNIGYRNQY-YQLZSBIMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nomegestrol acetate for ID and assay CRS EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Nomegestrol acetate (NOMAc) is a potent, highly selective progestogen, a full agonist at the progesterone receptor, with no or minimal binding to other steroid receptors. It shows potent antigonadotropic activity mediated at the hypothalamic and pituitary level with strong antiestrogenic effects and a partial antiandrogenic effect. NOMAc is used in various formulations as a contraceptive.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service