Skip to Content
MilliporeSigma

BP449

Progesterone

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

4-Pregnene-3,20-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
EC Number:
200-350-6
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
1915950
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

progesterone

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

128-132 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1

InChI key

RJKFOVLPORLFTN-LEKSSAKUSA-N

Gene Information

human ... PGR(5241)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Progesterone BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Progesterone Injection
  • Biochem/physiol Actions

    Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation. Progesterone is implicated in the etiology of breast cancer.
    Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation.

    Packaging

    Unit quantity: 50 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.
    Steroid hormone produced by the corpus luteum.


    Still not finding the right product?

    Explore all of our products under Progesterone


    pictograms

    Health hazard

    signalword

    Danger

    Hazard Classifications

    Carc. 2 - Lact. - Repr. 1A

    Storage Class

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    wgk

    WGK 3

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library