Key Documents
SU860098
Supelco
Headspace vial, screw top, rounded bottom (vial only)
volume 20 mL, amber glass vial, thread for 18, O.D. × H 22.5 mm × 75.5 mm, pkg of 100 ea
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
amber glass vial
round bottom
feature
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
22.5 mm × 75.5 mm
volume
20 mL
fitting
thread for 18
compatibility
for use with CTC PAL (Varian, Gerstel, Atas, Shimadzu)
General description
These vials require a special cap with an 18 mm thread. The standard 18-400 threaded caps will not fit this vial. The part numbers for caps that are available for these vials are SU860101(with PTFE/silicone, 1.3 mm thick), SU860102 (with PTFE/butyl, 1.6 mm thick), and SU860103 (with PTFE/silicone, 1.5 mm thick). These caps are made from stainless steel and are magnetic.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
High performance, reliability, and reproducibility of HS-SPME in combination with GC/MS for the determination of VOCs in water was proven in an interlaboratory trial. The new ISO 17943 using HS-SPME is an improvement on existing official methods for this determination in terms of sensitivity and selectivity.
High performance, reliability, and reproducibility of HS-SPME in combination with GC/MS for the determination of VOCs in water was proven in an interlaboratory trial. The new ISO 17943 using HS-SPME is an improvement on existing official methods for this determination in terms of sensitivity and selectivity.
High performance, reliability, and reproducibility of HS-SPME in combination with GC/MS for the determination of VOCs in water was proven in an interlaboratory trial. The new ISO 17943 using HS-SPME is an improvement on existing official methods for this determination in terms of sensitivity and selectivity.
High performance, reliability, and reproducibility of HS-SPME in combination with GC/MS for the determination of VOCs in water was proven in an interlaboratory trial. The new ISO 17943 using HS-SPME is an improvement on existing official methods for this determination in terms of sensitivity and selectivity.