Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL89180

Curcumin

phyproof® Reference Substance

Synonym(s):

(E,E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Diferuloylmethane, Diferulylmethane, Natural Yellow 3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOC6H3(OCH3)CH=CHCO]2CH2
CAS Number:
Molecular Weight:
368.38
Colour Index Number:
75300
Beilstein:
2306965
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

vapor density

13 (vs air)

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

form

powder

manufacturer/tradename

PhytoLab

SMILES string

COc1cc(\C=C\C(=O)CC(=O)\C=C\c2ccc(O)c(OC)c2)ccc1O

InChI

1S/C21H20O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h3-12,24-25H,13H2,1-2H3/b7-3+,8-4+

InChI key

VFLDPWHFBUODDF-FCXRPNKRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

A natural phenolic compound responsible for the yellow color of turmeric. Potent anti-tumor agent that acts against highly diverse tumor-specific pathways. Anti-inflammatory and anti-oxidant. Induces apoptosis in cancer cells and inhibits phorbol ester-induced protein kinase C (PKC) activity. Reported to inhibit production of inflammatory cytokines by peripheral blood monocytes and alveolar macrophages. Potent inhibitor of EGFR tyrosine kinase and IκB kinase. Inhibits inducible nitric oxide synthase (iNOS), cyclooxygenase (COX-2) and 5-lipoxygenase (5-LOX).
A natural phenolic compound. Potent anti-tumor agent having anti-inflammatory and anti-oxidant properties. Curcumin has been cited as a potential chemopreventive agent, in addition to its chemotherapeutic activity. Induces apoptosis in cancer cells and inhibits phorbol ester-induced protein kinase C (PKC) activity. Reported to inhibit production of inflammatory cytokines by peripheral blood monocytes and alveolar macrophages. Potent inhibitor of EGFR tyrosine kinase and IκB kinase. Inhibits inducible nitric oxide synthase (iNOS), cycloxygenase and lipoxygenase. Easily penetrates into the cytoplasm of cells, accumulating in membranous structures such as plasma membrane, endoplasmic reticulum and nuclear envelope.

Other Notes

This compound is commonly found in plants of the genus: curcuma

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service