Skip to Content
MilliporeSigma

239802

Curcumin

from Curcuma longa L., ≥80% (HPLC), 5-lipoxygenase inhibitor, solid

Synonym(s):

Curcumin, Curcuma longa L., 1,7-Bis(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, NOD2 Signaling Inhibitor I, Nucleotide-binding Oligomerization Domain 2 Signaling Inhibitor I, Histone Acetyltransferase Inhibitor I, HAT Inhibitor I, p300/CBP Inhibitor I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H20O6
CAS Number:
Molecular Weight:
368.38
UNSPSC Code:
12352205
NACRES:
NA.77
MDL number:
Assay:
≥80% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Curcumin, Curcuma longa L., A cell-permeable and irreversible antitumor and anti-inflammatory agent that acts as an inhibitor of 5-lipoxygenase (IC50 = 8 µM) and cyclooxygenase (IC50 = 52 µM).

Quality Segment

description

Merck USA index - 14, 2673

assay

≥80% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

orange-yellow

solubility

acetic acid: 10 mg/mL, DMSO: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C21H20O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h3-13,22,24-25H,1-2H3/b7-3+,8-4+,16-13-

InChI key

ZIUSSTSXXLLKKK-KOBPDPAPSA-N

General description

A cell permeable antitumor and anti-inflammatory agent that acts as an inhibitor of 5-lipoxygenase (IC50 = 8 µM) and cyclooxygenase (IC50 = 52 µM). Confers significant protection against neurotoxic and genotoxic agents. Also inhibits the induction of nitric oxide synthase in activated macrophages (IC50 = 6 µM). Recently shown to inhibit the EGF receptor intrinsic kinase activity in the human epidermoid carcinoma A431 cells in a dose- and time-dependent manner. Also shown to be a p300/CREB-binding protein-specific inhibitor of histone acetyltransferase, inhibiting the acetylation of histones H3 and H4 with an

IC50 of ~25 µM. Does not affect p300/CREB binding protein-associated factor (PCAF).
A cell-permeable and irreversible antitumor and anti-inflammatory agent that acts as an inhibitor of 5-lipoxygenase (IC50 = 8 µM) and cyclooxygenase (IC50 = 52 µM). Confers significant protection against neurotoxic and genotoxic agents. Also inhibits the induction of nitric oxide synthase in activated macrophages (IC50 = 6 µM). Recently shown to inhibit the EGF receptor intrinsic kinase activity in the human epidermoid carcinoma A431 cells in a dose- and time-dependent manner. Also shown to be a p300/CREB-binding protein-specific inhibitor of histone acetyltransferase, inhibiting the acetylation of histones H3 and H4 with an IC50 of ~25 µM. Does not affect p300/CREB binding protein-associated factor (PCAF).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
5-lipoxygenase
Product does not compete with ATP.
Reversible: no
Target IC50: 8 µM, 52 µM, against 5-lipoxygenase, Cyclooxygenase, respectively; 6 µM inhibiting the induction of nitric oxide synthase in activated macrophages

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hung, S., et al. 2008. Mol. Pharmacol.74, 274.
Cui, L., et al. 2007. Antimicrob. Agents Chemother.51, 488.
Salvioli, S., et al. 2007. eCAM4, 181.
Balasubramanyam, K. et al. 2004. J. Biol. Chem.279, 51163.
Brouet, I., and Okshima, H. 1995. Biochem. Biophys. Res. Commun. 206, 533.
Korutla, L., and Kumar, R. 1994. Biochim. Biophys. Acta1224, 597.
Flynn, D.L., et al. 1986. Prostagland. Leuk. Med. 22, 357.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under Curcumin, Curcuma longa L.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
239802-100MG04055977199581