Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHL89175

Chlorogenic acid

phyproof® Reference Substance

Synonym(s):

3-Caffeoylquinic acid (CAS), 5-Caffeoylquinic acid (IUPAC), 1,4,5-Trihydroxycyclohexanecarboxylic acid 3-(3,4-dihydroxycinnamate), 3-(3,4-Dihydroxycinnamoyl)quinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O9
CAS Number:
Molecular Weight:
354.31
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

mp

210 °C (dec.) (lit.)

SMILES string

O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@@H]1O)C(O)=O

InChI

1S/C16H18O9/c17-9-3-1-8(5-10(9)18)2-4-13(20)25-12-7-16(24,15(22)23)6-11(19)14(12)21/h1-5,11-12,14,17-19,21,24H,6-7H2,(H,22,23)/b4-2+/t11-,12-,14-,16+/m1/s1

InChI key

CWVRJTMFETXNAD-JUHZACGLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

  • Antioxidant and Anti-inflammatory Effects: Chlorogenic acid demonstrates significant protective effects against UV-B-induced skin damage through its antioxidant, anti-inflammatory, and anti-glycosylation properties, enhancing its potential for applications in dermatological health and cosmetic products (Wang et al., 2024).
  • Enhancement of Nutritional and Flavor Qualities in Agriculture: Studies have highlighted chlorogenic acid′s role in improving the nutritional and flavor qualities of tomatoes when exogenous silicon is applied, emphasizing its importance in agricultural food science (Yang et al., 2024).
  • Chemo-preventive and Therapeutic Potentials in Cancer: Chlorogenic acid is identified for its chemo-preventive and therapeutic capabilities in breast cancer management, providing substantial evidence for its application in oncology research (Olayiwola and Gollahon, 2024).

Other Notes

This compound is commonly found in plants of the genus: angelica arnica carum crataegus echinacea hedera humulus hypericum melissa pimpinella plantago salvia sambucus valeriana

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service