Skip to Content
MilliporeSigma
Chlorogenic acid European Pharmacopoeia (EP) Reference Standard

Chlorogenic acid

Chlorogenic acid European Pharmacopoeia (EP) Reference Standard
Synonym(s):
1,4,5-Trihydroxycyclohexanecarboxylic acid 3-(3,4-dihydroxycinnamate), 3-(3,4-Dihydroxycinnamoyl)quinic acid, Chlorogenic acid
Empirical Formula (Hill Notation):
C16H18O9
CAS Number:
Molecular Weight:
354.31
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique