Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL86198

Piperlongumine

phyproof® Reference Substance

Synonym(s):

5,6-Dihydro-1-(1-oxo-3-[3,4,5-trimethoxyphenyl]-trans-2-propenyl)-2[1H]-pyridinone, 5,6-Dihydro-1-[(2E)-1-oxo-3-(3,4,5-trimethoxyphenyl)-2-propen-1-yl]-2(1H)-Pyridinone, Piplartin, Piplartine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO5
CAS Number:
Molecular Weight:
317.34
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0%

manufacturer/tradename

PhytoLab

application(s)

food and beverages
pharmaceutical

storage temp.

2-8°C

SMILES string

COc1cc(\C=C\C(=O)N2CCC=CC2=O)cc(OC)c1OC

InChI

1S/C17H19NO5/c1-21-13-10-12(11-14(22-2)17(13)23-3)7-8-16(20)18-9-5-4-6-15(18)19/h4,6-8,10-11H,5,9H2,1-3H3/b8-7+

InChI key

VABYUUZNAVQNPG-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Piperlongumine selectively kills cancer cells regardless of p53 status without harming normal cells. It binds to and inihbits proteins known to regulate oxidative stress, in particular, Glutathione S-transferase pi 1 (GSTP1). It increases the level of reactive oxygen species (ROS) and apoptotic cell death in cancer cells with little effect on either rapidly or slowly dividing primary normal cells. Piperlongumine showed significant antitumour effects in a variety of mouse tumour models and inhibited growth of spontaneously formed malignant breast tumours and their associated metastases.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service