Skip to Content
MilliporeSigma

528124

Piperlongumine

A cell-permeable, orally bioavailable natural product isolated from the plant species, Piper longum L, which selectively induces cell death in cancer cells that express both wild-type and mutant p53.

Synonym(s):

Piperlongumine, Piplartine, (E)-1-(3-(3,4,5-Trimethoxyphenyl)acryloyl)-5,6-dihydropyridin-2(1H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H19NO5
CAS Number:
Molecular Weight:
317.34
UNSPSC Code:
12352100
NACRES:
NA.77
MDL number:
Assay:
≥97% (HPLC)
Form:
solid
Quality level:
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1N(C(/C=C/C2=CC(OC)=C(OC)C(OC)=C2)=O)CCC=C1

InChI

1S/C17H19NO5/c1-21-13-10-12(11-14(22-2)17(13)23-3)7-8-16(20)18-9-5-4-6-15(18)19/h4,6-8,10-11H,5,9H2,1-3H3/b8-7+

InChI key

VABYUUZNAVQNPG-BQYQJAHWSA-N

General description

A cell-permeable, orally bioavailable natural product isolated from the plant species, Piper longum L, which selectively induces cell death in cancer cells that express both wild-type and mutant p53, dose-dependently from 1–15 µM, but not in primary normal cells and non-transformed immortalized cells. It significantly enhances wild-type p53 and PUMA expression, and inhibits the expression of several pro-survival proteins, including BCL2, surviving, and XIAP, in various types of cancer cells, but not in normal cells. It elicits ROS and apoptosis in several human cancer cells (EJ, MDA-MB-231, U2OS and MDA-MB-435), possibly through interaction with oxidative stress responders such as GSTP1 and CBR1, and can be reversed by NAC (3 mM) in EJ cells. Additionally, it exhibits anti-tumour effects and inhibits the formation of blood vessels in xenograft-tumour mice without obvious toxicity, and inhibits tumor growth and their associated metastases in a MMTV-PyVT breast cancer transgenic mouse model ( 2.4 mg/kg, daily for two weeks, i.p.), which is more effective than paclitaxel.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Raj, L., et al. 2011. Nature475, 231.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Regulatory Review (Z)


Still not finding the right product?

Explore all of our products under Piperlongumine


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
528124-25MG04055977196382