PHL84241
Mogrol
phyproof® Reference Substance
Synonym(s):
(3β,9β,10α,11α,24R)-9-Methyl-19-norlanost-5-ene-3,11,24,25-tetrol
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥90.0% (HPLC)
form
solid
manufacturer/tradename
PhytoLab
SMILES string
O[C@H]1[C@@]2([C@H]([C@]4([C@@]([C@H](CC4)[C@@H](CC[C@@H](O)C(O)(C)C)C)(C1)C)C)CC=C3[C@H]2CC[C@@H](C3(C)C)O)C
InChI
1S/C30H52O4/c1-18(9-13-24(32)27(4,5)34)19-15-16-28(6)22-12-10-20-21(11-14-23(31)26(20,2)3)30(22,8)25(33)17-29(19,28)7/h10,18-19,21-25,31-34H,9,11-17H2,1-8H3/t18-,19-,21-,22+,23+,24-,25-,28+,29-,30+/m1/s1
InChI key
JLYBBRAAICDTIS-AYEHCKLZSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Biochem/physiol Actions
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service