Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL83333

Decursin

phyproof® Reference Substance

Synonym(s):

(+)-Decursin, (7S)-7,8-Dihydro-8,8-dimethyl-2-oxo-2H,6H-benzo[1,2-b:5,4-b′]dipyran-7-yl 3-methyl-2-butenoate, 3-Methyl-2-butenoic acid (7S)-7,8-dihydro-8,8-dimethyl-2-oxo-2H,6H-pyrano[3,2-g]-1-benzopyran-7-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20O5
CAS Number:
Molecular Weight:
328.36
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0%

manufacturer/tradename

PhytoLab

application(s)

food and beverages
pharmaceutical

storage temp.

2-8°C

SMILES string

[o]1c2c(cc3c(c2)OC([C@H](C3)OC(=O)C=C(C)C)(C)C)cc[c]1=O

InChI

1S/C19H20O5/c1-11(2)7-18(21)23-16-9-13-8-12-5-6-17(20)22-14(12)10-15(13)24-19(16,3)4/h5-8,10,16H,9H2,1-4H3/t16-/m0/s1

InChI key

CUKSFECWKQBVED-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Decursin is a cumarin isolated from Radix peucedani that exhibits a number of physiological actions including anticancer, antibacterial, antinematodal and antioxidant.
Decursin is a cumarin isolated from Radix peucedani that exhibits a number of physiological actions including anticancer, antibacterial, antinematodal and antioxidant. Decursin is potent anti-angiogenic agent targeting the VEGFR-2 signaling pathway. Decursin exhibits potent neuroprotective activity against glutamate and Ab- induced neurotoxicity. Apparently, decursin stimulates activation of Nerf2. Also, decursin inhibits androgen stimulated AR translocation to the nucleus in LNCaP prostate cancer cells.

Other Notes

This compound is commonly found in plants of the genus: angelica

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service