Skip to Content
MilliporeSigma

SML3329

Decursinol Angelate

≥98% (HPLC), PKC activator, powder

Synonym(s):

(2Z)​-2-​Methyl-2-​butenoic acid (7S)​-​7,​8-​dihydro-​8,​8-​dimethyl-​2-​oxo-​2H,​6H-​benzo[1,​2-​b:5,​4-​b′]​dipyran-​7-​yl ester, Acutilobin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H20O5
CAS Number:
Molecular Weight:
328.36
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Form:
powder
Assay:
≥98% (HPLC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Decursinol Angelate, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[o]1c2c(cc3c(c2)OC([C@H](C3)OC(=O)\C(=C/C)\C)(C)C)cc[c]1=O

InChI

1S/C19H20O5/c1-5-11(2)18(21)23-16-9-13-8-12-6-7-17(20)22-14(12)10-15(13)24-19(16,3)4/h5-8,10,16H,9H2,1-4H3/b11-5-/t16-/m0/s1

InChI key

AGABNGOXUSXQDD-XKGFZTIGSA-N

Biochem/physiol Actions

Decursinol Angelate is a pyranocoumarin isolated from medical plant Angelica gigas that exhibits potent anti-inflammatory properties. Decursinol Angelate appears to suppress type 17 helper T (Th17) cell responses in dextran sodium sulfate-induced colitis model. Decursinol Angelate reduces the production of NF-κB and COX-2 induced proinflammatory cytokine levels on TPA induced mice ear edema.
Main anti-inflammatory component of Angelica gigas extract


Still not finding the right product?

Explore all of our products under Decursinol Angelate


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library