Skip to Content
Merck
All Photos(1)

Key Documents

G006129

HPLC Analysis of E. coli Lysate on Proteomix® SAX Columns, Particle Size Comparison

application for HPLC

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item


technique(s)

HPLC: suitable

test parameters

column temp.: 25 °C
column: Proteomix SAX-NP10, 5 cm x 4.6 mm I.D., 10 μm particles (Z777203)
column: Proteomix SAX-NP5, 5 cm x 4.6mm I.D., 5 μm particles (Z777227)
detector: UV, 280 nm
flow rate: 0.5 mL/min
gradient: 0 - 100%B in 30 min
injection: 10 μL
mobile phase: (A) 20 mM Tris, pH 9.0; (B) A + 0.5 M NaCl
sample: 2.5 mg/mL

suitability

application for HPLC

application(s)

life science and biopharma
pharmaceutical (small molecule)

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service