Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM78266

Supelco

Isovaleric acid

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3-Methylbutanoic acid, 3-Methylbutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COOH
CAS Number:
Molecular Weight:
102.13
Beilstein:
1098522
MDL number:
UNSPSC Code:
41116107

grade

certified reference material
TraceCERT®

Quality Level

vapor pressure

0.38 mmHg ( 20 °C)

product line

TraceCERT®

autoignition temp.

824 °F

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n20/D 1.403 (lit.)

bp

175-177 °C (lit.)

mp

−29 °C (lit.)

density

0.925 g/mL at 20 °C (lit.)

SMILES string

CC(C)CC(O)=O

InChI

1S/C5H10O2/c1-4(2)3-5(6)7/h4H,3H2,1-2H3,(H,6,7)

InChI key

GWYFCOCPABKNJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

176.0 °F - Pensky-Martens closed cup

Flash Point(C)

80 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service