Skip to Content
MilliporeSigma
Isovaleric acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Isovaleric acid

Isovaleric acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
3-Methylbutanoic acid, 3-Methylbutyric acid
Linear Formula:
(CH3)2CHCH2COOH
CAS Number:
Molecular Weight:
102.13

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique