Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRM42356

Supelco

Sodium diethyldithiocarbamate trihydrate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Cupral, Diethyldithiocarbamic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCSSNa · 3H2O
CAS Number:
Molecular Weight:
225.31
Beilstein:
3920507
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

5.9 (vs air)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

mp

95-98.5 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]O[H].[H]O[H].[H]O[H].CCN(CC)C(=S)S[Na]

InChI

1S/C5H11NS2.Na.3H2O/c1-3-6(4-2)5(7)8;;;;/h3-4H2,1-2H3,(H,7,8);;3*1H2/q;+1;;;/p-1

InChI key

WWGXHTXOZKVJDN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO2 has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

Biochem/physiol Actions

Inhibits induction of macrophage nitric oxide synthase.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service