Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.06689

Supelco

Sodium diethyldithiocarbamate trihydrate

(reagent for copper) GR for analysis ACS

Synonym(s):

Sodium diethyldithiocarbamate trihydrate, Diethyldithiocarbamic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10NNaS2 · 3H2O
CAS Number:
Molecular Weight:
225.31
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-710-6
NACRES:
NA.05

grade

ACS reagent

Quality Level

form

solid

potency

1500 mg/kg LD50, oral (Rat)
>1000 mg/kg LD50, skin (Rat)

specific analyte(s)

sodium

pH

7 (20 °C, 20 g/L in H2O)

mp

93 °C

solubility

600 g/L

storage temp.

2-8°C

InChI

1S/C5H11NS2.Na.3H2O/c1-3-6(4-2)5(7)8;;;;/h3-4H2,1-2H3,(H,7,8);;3*1H2/q;+1;;;/p-1

InChI key

WWGXHTXOZKVJDN-UHFFFAOYSA-M

Analysis Note

Assay (HClO₄): ≥ 97.0 %
Sulfated ash: 30.0 - 36.0 %
Sensitivity (as reagent of copper): passes test
Identity (IR): passes test
Corresponds to ACS

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination with PAR subsequent to separation of lead with Sodium diethyldithiocarbaminate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service