Skip to Content
MilliporeSigma
All Photos(1)

Documents

BP1263

Tranexamic acid

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

trans-4-(Aminomethyl)cyclohexanecarboxylic acid, AMCA, AMCHA, HAKU, TAMCHA, Tranexamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2C6H10CO2H
CAS Number:
Molecular Weight:
157.21
Beilstein:
2207452
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tranexamic acid

form

crystalline

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

>300 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

NC[C@H]1CC[C@@H](CC1)C(O)=O

InChI

1S/C8H15NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h6-7H,1-5,9H2,(H,10,11)/t6-,7-

InChI key

GYDJEQRTZSCIOI-LJGSYFOKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tranexamic acid BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Tranexamic Acid Injection
  • Tranexamic Acid Oral Solution
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Pictograms

    Exclamation mark

    Signal Word

    Warning

    Hazard Statements

    Hazard Classifications

    Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

    Target Organs

    Respiratory system

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 2

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service