Skip to Content
MilliporeSigma
Tranexamic acid European Pharmacopoeia (EP) Reference Standard

Tranexamic acid

Tranexamic acid European Pharmacopoeia (EP) Reference Standard
Synonym(s):
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, AMCA, AMCHA, HAKU, TAMCHA, Tranexamic acid
Linear Formula:
H2NCH2C6H10CO2H
CAS Number:
Molecular Weight:
157.21

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique