Skip to Content
MilliporeSigma
All Photos(1)

Documents

94056

Supelco

1,3,5-Tri-tert-butylbenzene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3[C(CH3)3]3
CAS Number:
Molecular Weight:
246.43
Beilstein:
1909579
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

form

crystals

shelf life

limited shelf life, expiry date on the label

bp

121-122 °C/12 mmHg (lit.)

mp

67-72 °C (lit.)

SMILES string

CC(C)(C)c1cc(cc(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C18H30/c1-16(2,3)13-10-14(17(4,5)6)12-15(11-13)18(7,8)9/h10-12H,1-9H3

InChI key

GUFMBISUSZUUCB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The 1,3,5-Tri-tert-butylbenzene may find the following uses:

  • The qualitative study of pyrolytic product vapor of Helianthus annuus stems using pyrolysis–gas chromatography-mass spectrometry (Py-GC/MS)
  • Gas-phase synthesis and Fourier-transform ion cyclotron resonance (FT/ICR) mass spectrometric characterization of sandwich complexes of Sc+, Y+, and lanthanide ions with 1,3,5 -tri-tert-butylbenzene

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service